Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- LTI78
- Entrez Gene ID
-
835307
- Gene Name
- low-temperature-responsive protein 78 (LTI78) / desiccation-responsive protein 29A (RD29A)
- Gene Aliases
-
AT5G52310, COLD REGULATED 78, COR78, K24M7.4, K24M7_4, LOW-TEMPERATURE-INDUCED 78, LTI140, RD29A, RESPONSIVE TO DESSICATION 29A
- Chromosome Location
-
Chr.5: 21240717 - 21243644 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124610.3 |
NP_200044.1 |
1 - 2 |
263 |
67 |
GenBank mRNA |
L22567.1 |
- |
1 - 2 |
129 |
67 |
View Full Product Details