Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ML2
Entrez Gene ID
818890
Gene Name
MEI2-like 2
Gene Aliases
AML2, AT2G42890, F7D19.11, F7D19_11, MEI2-like 2
Chromosome Location
Chr.2: 17849819 - 17854553 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084574.1 NP_001078043.1 12 - 13 2533 80
NM_129850.4 NP_565990.1 12 - 13 2664 80
NM_201945.3 NP_973674.1 11 - 12 2707 80
GenBank mRNA AF367316.1 - 12 - 13 2490 80
AY143940.1 - 12 - 13 2406 80

View Full Product Details