Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARAB-1
Entrez Gene ID
839767
Gene Name
GDSL-like Lipase/Acylhydrolase superfamily protein
Gene Aliases
AT1G28670
Chromosome Location
Chr.1: 10074359 - 10076457 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_102634.3 NP_174188.1 1 - 2 474 88
GenBank mRNA AK118766.1 - 1 - 2 361 88
U38916.1 - 1 - 2 276 88

View Full Product Details