Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
STV1
Entrez Gene ID
824468
Gene Name
Ribosomal protein L24e family protein
Gene Aliases
AT3G53020, RIBOSOMAL PROTEIN L24, RPL24, RPL24B, SHORT VALVE1
Chromosome Location
Chr.3: 19660488 - 19662025 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_115162.4 NP_190870.1 4 - 5 451 174
GenBank mRNA AB199790.1 - 4 - 5 386 174
AY058129.1 - 4 - 5 451 174
AY093180.1 - 4 - 5 405 174
BT006545.1 - 3 - 4 338 174

View Full Product Details