Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PCL1
Entrez Gene ID
823817
Gene Name
Homeodomain-like superfamily protein
Gene Aliases
AT3G46640, LUX, LUX ARRHYTHMO, PHYTOCLOCK 1
Chromosome Location
Chr.3: 17183042 - 17186959 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035746.2 NP_001030823.1 1 - 2 1171 111

View Full Product Details