Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM53C
Entrez Gene ID
534006
Gene Name
family with sequence similarity 53 member C
Gene Aliases
-
Chromosome Location
Chr.7: 49736535 - 49746612 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001046366.1 NP_001039831.1 2 - 3 272 77
GenBank mRNA BC114114.1 - 2 - 3 272 77

View Full Product Details