Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBLN5
Entrez Gene ID
535185
Gene Name
fibulin 5
Gene Aliases
-
Chromosome Location
Chr.21: 56567084 - 56659763 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014946.1 NP_001014946.1 6 - 7 995 64
GenBank mRNA BC105467.1 - 6 - 7 984 64
BT020707.1 - 6 - 7 995 64

View Full Product Details