Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IBSP
Entrez Gene ID
281233
Gene Name
integrin binding sialoprotein
Gene Aliases
-
Chromosome Location
Chr.6: 36879556 - 36893855 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174084.3 NP_776509.1 1 - 2 32 89
GenBank mRNA S73144.1 - 1 - 2 36 89

View Full Product Details