Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PDE6D
Entrez Gene ID
281976
Gene Name
phosphodiesterase 6D
Gene Aliases
-
Chromosome Location
Chr.2: 119443675 - 119498744 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174420.2 NP_776845.1 2 - 3 307 92
GenBank mRNA U65073.1 - 2 - 3 307 92

View Full Product Details