Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAFAH1B3
Entrez Gene ID
282515
Gene Name
platelet activating factor acetylhydrolase 1b catalytic subunit 3
Gene Aliases
-
Chromosome Location
Chr.18: 50978724 - 50981557 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174665.2 NP_777090.1 1 - 2 55 81
GenBank mRNA BC120202.1 - 1 - 2 80 81
D30789.1 - 1 - 2 55 81

View Full Product Details