Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PAFAH1B3
Entrez Gene ID
282515
Gene Name
platelet activating factor acetylhydrolase 1b catalytic subunit 3
Gene Aliases
-
Chromosome Location
Chr.18: 50978724 - 50981557 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174665.2 NP_777090.1 4 - 5 357 71
GenBank mRNA BC120202.1 - 4 - 5 382 71
D30789.1 - 4 - 5 357 71

View Full Product Details