Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- CATHL4
- Entrez Gene ID
-
282166
- Gene Name
- cathelicidin 4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 51592817 - 51594189 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174827.2 |
NP_777252.1 |
1 - 2 |
209 |
59 |
GenBank mRNA |
X67340.1 |
- |
- |
209 |
59 |
- Gene Symbol
- LOC112443481
- Entrez Gene ID
-
112443481
- Gene Name
- cathelicidin-4
- Gene Aliases
-
-
- Chromosome Location
-
Chr.22: 51586118 - 51587494 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC133480.1 |
- |
1 - 2 |
214 |
59 |
View Full Product Details