Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMA5
Entrez Gene ID
510155
Gene Name
proteasome 20S subunit alpha 5
Gene Aliases
-
Chromosome Location
Chr.3: 34012638 - 34033663 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015566.1 NP_001015566.1 7 - 8 657 109
GenBank mRNA BC102343.1 - 7 - 8 650 109
BT021033.1 - 7 - 8 657 109

View Full Product Details