Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BHLHE40
Entrez Gene ID
506945
Gene Name
basic helix-loop-helix family member e40
Gene Aliases
BHLHB2
Chromosome Location
Chr.22: 21400616 - 21406318 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001024929.1 NP_001020100.1 4 - 5 611 125
GenBank mRNA BT020754.1 - 4 - 5 611 125

View Full Product Details