Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MARCHF5
Entrez Gene ID
540937
Gene Name
membrane associated ring-CH-type finger 5
Gene Aliases
MARCH5
Chromosome Location
Chr.26: 13778743 - 13829856 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034715.2 NP_001029887.1 3 - 4 688 84
GenBank mRNA BC102966.1 - 3 - 4 690 84

View Full Product Details