Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTLL3
Entrez Gene ID
531866
Gene Name
tubulin tyrosine ligase-like family, member 3
Gene Aliases
-
Chromosome Location
Chr.22: 16888517 - 16911633 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BT021865.1 - 6 - 7 1295 59

View Full Product Details