Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MYF6
Entrez Gene ID
281336
Gene Name
myogenic factor 6
Gene Aliases
MRF4
Chromosome Location
Chr.5: 10275399 - 10277216 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_181811.2 NP_861527.1 2 - 3 692 84
GenBank mRNA AB110601.1 - 2 - 3 613 84
AB257561.1 - 2 - 3 640 84
BC142159.1 - 2 - 3 692 84

View Full Product Details