Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CNBP
Entrez Gene ID
504831
Gene Name
CCHC-type zinc finger nucleic acid binding protein
Gene Aliases
-
Chromosome Location
Chr.22: 59070493 - 59080254 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034224.1 NP_001029396.1 2 - 3 237 76
GenBank mRNA BC102298.1 - 2 - 3 237 76

View Full Product Details