Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BET1L
Entrez Gene ID
509758
Gene Name
Bet1 golgi vesicular membrane trafficking protein like
Gene Aliases
-
Chromosome Location
Chr.11: 82867454 - 82873253 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034362.2 NP_001029534.1 2 - 3 173 77
GenBank mRNA BC105155.1 - 2 - 3 174 77

View Full Product Details