Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PRRG2
Entrez Gene ID
511235
Gene Name
proline rich and Gla domain 2
Gene Aliases
-
Chromosome Location
Chr.18: 56002171 - 56008470 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034402.2 NP_001029574.1 1 - 2 140 89
GenBank mRNA BC102320.1 - 1 - 2 142 89

View Full Product Details