Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BDH2
Entrez Gene ID
515321
Gene Name
3-hydroxybutyrate dehydrogenase 2
Gene Aliases
DHRS6
Chromosome Location
Chr.6: 21762056 - 21790795 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034488.2 NP_001029660.1 3 - 4 329 98
GenBank mRNA BC102567.1 - 3 - 4 330 98

View Full Product Details