Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LMO4
Entrez Gene ID
614212
Gene Name
LIM domain only 4
Gene Aliases
-
Chromosome Location
Chr.3: 56698567 - 56715683 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034751.1 NP_001029923.1 3 - 4 393 67
GenBank mRNA BC104582.1 - 3 - 4 393 67

View Full Product Details