Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MISP
Entrez Gene ID
505316
Gene Name
mitotic spindle positioning
Gene Aliases
C7H19orf21
Chromosome Location
Chr.7: 43324443 - 43335563 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035269.2 NP_001030346.1 4 - 5 2126 117
GenBank mRNA BC107524.1 - 4 - 5 2128 117

View Full Product Details