Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
URI1
Entrez Gene ID
515119
Gene Name
URI1 prefoldin like chaperone
Gene Aliases
C18H19orf2, RMP
Chromosome Location
Chr.18: 40542851 - 40608332 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035305.2 NP_001030382.1 1 - 2 410 149
GenBank mRNA BC107540.1 - 1 - 2 411 149
BT030747.1 - 1 - 2 303 149

View Full Product Details