Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL5
Entrez Gene ID
515238
Gene Name
ribosomal protein L5
Gene Aliases
-
Chromosome Location
Chr.3: 50629955 - 50638559 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035306.2 NP_001030383.1 1 - 1 598 89
GenBank mRNA BC105198.1 - 1 - 1 599 89
BT021482.1 - 5 - 6 575 89

View Full Product Details