Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC617486
Entrez Gene ID
617486
Gene Name
chromosome 3 open reading frame, human C1orf88-like
Gene Aliases
C1orf88, C3H1orf88
Chromosome Location
Chr.3: 31952315 - 31958001 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076464.2 NP_001069932.1 1 - 2 63 101
GenBank mRNA BC109727.1 - 1 - 2 64 101

View Full Product Details