Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AQP7
Entrez Gene ID
615498
Gene Name
aquaporin 7
Gene Aliases
-
Chromosome Location
Chr.8: 75135985 - 75154407 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076378.2 NP_001069846.1 1 - 2 237 83
GenBank mRNA BC109728.1 - 1 - 2 238 83

View Full Product Details