Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC2HC1B
Entrez Gene ID
768003
Gene Name
zinc finger C2HC-type containing 1B
Gene Aliases
C6orf94, C9H6orf94, FAM164B
Chromosome Location
Chr.9: 81316649 - 81370619 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077063.2 NP_001070531.1 7 - 8 714 103
GenBank mRNA BC110003.1 - 7 - 8 715 103

View Full Product Details