Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IL4R
Entrez Gene ID
404154
Gene Name
interleukin 4 receptor
Gene Aliases
-
Chromosome Location
Chr.25: 24898783 - 24922680 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075142.1 NP_001068610.1 9 - 10 956 61
GenBank mRNA BC118151.1 - 9 - 10 956 61

View Full Product Details