Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NAF1
Entrez Gene ID
514182
Gene Name
nuclear assembly factor 1 ribonucleoprotein
Gene Aliases
-
Chromosome Location
Chr.6: 2620826 - 2671246 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075652.2 NP_001069120.2 2 - 3 564 134
GenBank mRNA BC114778.1 - 2 - 3 565 134
DV822960.1 - 2 - 3 491 134
DV845783.1 - 2 - 3 565 134

View Full Product Details