Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GOLPH3
Entrez Gene ID
531359
Gene Name
golgi phosphoprotein 3
Gene Aliases
-
Chromosome Location
Chr.20: 41439630 - 41491579 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075948.1 NP_001069416.1 3 - 4 777 88
GenBank mRNA BC114663.1 - 3 - 4 777 88

View Full Product Details