Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN3
Entrez Gene ID
511251
Gene Name
neuroligin 3
Gene Aliases
-
Chromosome Location
Chr.X: 79470571 - 79493211 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075504.1 NP_001068972.1 2 - 3 748 63
GenBank mRNA BC123785.1 - 2 - 3 748 63

View Full Product Details