Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALDH8A1
Entrez Gene ID
513537
Gene Name
aldehyde dehydrogenase 8 family member A1
Gene Aliases
-
Chromosome Location
Chr.9: 72977326 - 73001933 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075626.1 NP_001069094.1 3 - 4 469 102
GenBank mRNA BC120092.1 - 3 - 4 469 102
BT029881.1 - 3 - 4 480 102

View Full Product Details