Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TM4SF1
Entrez Gene ID
533038
Gene Name
transmembrane 4 L six family member 1
Gene Aliases
-
Chromosome Location
Chr.1: 118809503 - 118817969 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075980.1 NP_001069448.1 4 - 5 682 91
GenBank mRNA BC119952.1 - 4 - 5 682 91
BT029905.1 - 4 - 5 673 91

View Full Product Details