Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZMAT4
Entrez Gene ID
616397
Gene Name
zinc finger matrin-type 4
Gene Aliases
-
Chromosome Location
Chr.27: 35579521 - 35955896 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076943.1 NP_001070411.1 6 - 7 803 102
GenBank mRNA BC119856.1 - 6 - 7 803 102

View Full Product Details