Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LXN
Entrez Gene ID
781644
Gene Name
latexin
Gene Aliases
-
Chromosome Location
Chr.1: 108867031 - 108874757 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080340.2 NP_001073809.1 4 - 5 864 149
GenBank mRNA BC120149.1 - 4 - 5 865 149

View Full Product Details