Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARRDC1
Entrez Gene ID
786098
Gene Name
arrestin domain containing 1
Gene Aliases
-
Chromosome Location
Chr.11: 106567613 - 106575152 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080365.2 NP_001073834.1 3 - 4 333 63
GenBank mRNA BC120279.1 - 3 - 4 334 63

View Full Product Details