Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
B2M
Entrez Gene ID
280729
Gene Name
beta-2-microglobulin
Gene Aliases
-
Chromosome Location
Chr.10: 103095452 - 103110775 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173893.3 NP_776318.1 2 - 3 383 142
GenBank mRNA AB098926.1 - 1 - 2 271 141
BC118352.1 - 2 - 3 383 142

Gene Symbol
LOC783680
Entrez Gene ID
783680
Gene Name
beta-2-microglobulin
Gene Aliases
-
Chromosome Location
Chr.10: 103116413 - 103122698 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA X69084.1 - - 356 142

View Full Product Details