Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
THEM4
Entrez Gene ID
787270
Gene Name
thioesterase superfamily member 4
Gene Aliases
-
Chromosome Location
Chr.3: 18844260 - 18893497 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080368.1 NP_001073837.1 4 - 5 563 144
GenBank mRNA BC126654.1 - 4 - 5 563 144

View Full Product Details