Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO5
Entrez Gene ID
515080
Gene Name
F-box protein 5
Gene Aliases
-
Chromosome Location
Chr.9: 89878600 - 89889345 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001082435.2 NP_001075904.1 3 - 4 1003 103
GenBank mRNA BC133649.1 - 3 - 4 1004 103

View Full Product Details