Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CIDEA
Entrez Gene ID
527051
Gene Name
cell death inducing DFFA like effector a
Gene Aliases
-
Chromosome Location
Chr.24: 42778433 - 42792110 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083449.1 NP_001076918.1 2 - 3 223 60
GenBank mRNA BC123394.1 - 2 - 3 223 60

View Full Product Details