Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SNRPE
Entrez Gene ID
531493
Gene Name
small nuclear ribonucleoprotein polypeptide E
Gene Aliases
-
Chromosome Location
Chr.16: 1636025 - 1641981 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083459.2 NP_001076928.1 1 - 1 260 109
GenBank mRNA BC114915.1 - 1 - 1 262 109

View Full Product Details