Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO47
Entrez Gene ID
613655
Gene Name
F-box protein 47
Gene Aliases
-
Chromosome Location
Chr.19: 39503233 - 39531332 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001083759.1 NP_001077228.1 8 - 9 1109 72
GenBank mRNA BC134684.1 - 8 - 9 1109 72

View Full Product Details