Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TNFRSF11B
Entrez Gene ID
523822
Gene Name
TNF receptor superfamily member 11b
Gene Aliases
-
Chromosome Location
Chr.14: 45276997 - 45305174 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001098056.2 NP_001091525.1 1 - 2 234 76
GenBank mRNA BC140651.1 - 1 - 2 235 76

View Full Product Details