Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
OLFML2B
Entrez Gene ID
513053
Gene Name
olfactomedin like 2B
Gene Aliases
-
Chromosome Location
Chr.3: 7530682 - 7580216 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100354.1 NP_001093824.1 3 - 4 587 98
GenBank mRNA BC147922.1 - 3 - 4 587 98

View Full Product Details