Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GPD1L
Entrez Gene ID
514456
Gene Name
glycerol-3-phosphate dehydrogenase 1 like
Gene Aliases
-
Chromosome Location
Chr.22: 6593957 - 6655425 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100363.2 NP_001093833.1 7 - 8 1051 80
GenBank mRNA BC149966.1 - 7 - 8 1052 80

View Full Product Details