Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
KRT23
Entrez Gene ID
510914
Gene Name
keratin 23
Gene Aliases
-
Chromosome Location
Chr.19: 41116546 - 41134104 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001101935.1 NP_001095405.1 6 - 7 1187 103
GenBank mRNA BC149166.1 - 6 - 7 1187 103

View Full Product Details