Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PYROXD1
Entrez Gene ID
505859
Gene Name
pyridine nucleotide-disulphide oxidoreductase domain 1
Gene Aliases
-
Chromosome Location
Chr.5: 88729719 - 88756464 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105334.1 NP_001098804.1 1 - 2 91 83
GenBank mRNA BC123726.1 - 1 - 2 91 83

View Full Product Details