Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FHL1
Entrez Gene ID
509056
Gene Name
four and a half LIM domains 1
Gene Aliases
-
Chromosome Location
Chr.Unknown: 8690290 - 8755071 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113258.1 NP_001106729.1 4 - 5 618 62
NM_001113259.1 NP_001106730.1 5 - 6 697 62
GenBank mRNA DV890085.1 - 4 - 5 569 62
DV931938.1 - 4 - 5 597 62
EE390515.1 - 4 - 5 620 62

View Full Product Details