Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CIB4
Entrez Gene ID
617324
Gene Name
calcium and integrin binding family member 4
Gene Aliases
-
Chromosome Location
Chr.11: 72879341 - 72983050 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076455.2 NP_001069923.2 4 - 5 383 78
GenBank mRNA BC109857.1 - 4 - 5 371 78
DT834350.1 - 4 - 5 384 78

View Full Product Details